Learn More
ACROBiosystems Cynomolgus SEZ6 100ug
Supplier:  ACROBiosystems SE6C52H4100UG
Encompass_Preferred
Cynomolgus SEZ6 Protein His Tag (SE6-C52H4) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - His 927 (Accession # A0A2K5WPJ4-1).
Catalog No. 50-253-4191
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.