Learn More
ACROBiosystems Cynomolgus Siglec-10 25ug
Supplier:  ACROBiosystems SI0C82E525UG
Encompass_Preferred
Biotinylated Cynomolgus Siglec-10 Protein HisAvitag (SI0-C82E5) is expressed from human 293 cells (HEK293). It contains AA Thr 17 - Ala 549 (Accession # XP_005590164.1).
Catalog No. 50-253-4180
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.