Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP1A1/CYP1A2 Monoclonal Antibody (MC1)
GREENER_CHOICE

Catalog No. 01670132 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-132 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-132 Supplier Invitrogen™ Supplier No. MA3036
  • $602.55
    $481.65 / Each of 1

    Save $120.90

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

MA3-036 detects Cytochrome P450 1A1/1A2 from human, primate, rat and mouse samples. MA3-036 has been successfully used in Western blot, immunocytochemistry, immunofluorescence and immunohistochemisty (paraffin) applications. By Western blot, this antibody detects a ~58 kDa band representing Cytochrome P450 1A1/1A2. The MA3-036 immunogen is 3-methylcholanthrene induced rat cytochrome P450 protein.

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. The protein encoded by this gene localizes to the endoplasmic reticulum and its expression is induced by some polycyclic aromatic hydrocarbons (PAHs), some of which are found in cigarette smoke. The enzyme's endogenous substrate is unknown; however, it is able to metabolize some PAHs to carcinogenic intermediates. Other xenobiotic substrates for this enzyme include caffeine, aflatoxin B1, and acetaminophen. The transcript from this gene contains four Alu sequences flanked by direct repeats in the 3' untranslated region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP1A1/CYP1A2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone MC1
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.05% sodium azide
Gene CYP1A2
Gene Accession No. P00184, P00185, P00186, P04798, P04799, P05177
Gene Alias AHH; AHRR; aromatic compound inducible; aryl hydrocarbon hydroxylase; cholesterol 25-hydroxylase; CP11; CP12; CYP1; Cyp1a1; Cyp1a-1; CYP1A2; Cyp1a-2; Cyp45c; Cypc45c; CYPD45; CYPIA1; CYPIA2; Cytochrome P(3)450; cytochrome P1-450; cytochrome P1-450, dioxin-inducible; Cytochrome P3 450; cytochrome P-448; cytochrome P450 1A1; Cytochrome P450 1A2; cytochrome P450 4; cytochrome P450 family 1 subfamily A member 1; cytochrome P450 family 1 subfamily A member 2; cytochrome P450 family 1 subfamily a polypeptide 1; cytochrome P450 family 1 subfamily a polypeptide 2; cytochrome P450 form 6; cytochrome P450 subfamily I (aromatic compound-inducible) member A1 (C6 form c) (C6 form c); Cytochrome P450 subfamily I (aromatic compound-inducible) member A2 (Q42 form d); cytochrome P450 subfamily I, polypeptide 1; cytochrome P450, 1a1; cytochrome P450, 1a1, aromatic compound inducible; cytochrome P450, 1a2; cytochrome P450, 1a2, aromatic compound inducible; cytochrome P450, family 1, subfamily a, polypeptide 1; cytochrome P450, family 1, subfamily a, polypeptide 2; cytochrome P450, subfamily I (aromatic compound-inducible), member A1 (C6, form c) (C6, form c); cytochrome P450, subfamily I (aromatic compound-inducible), member A2 (Q42, form d); cytochrome P450, subfamily I (aromatic compound-inducible), polypeptide 1; cytochrome P450, subfamily I (aromatic compound-inducible), polypeptide 2; cytochrome P450-C; Cytochrome P-450d; Cytochrome P450-D; Cytochrome P450MT2; Cytochrome P450MT2A; Cytochrome P450MT2B; cytochrome P450-P1; Cytochrome P450-P2; cytochrome P450-P3; dioxin-inducible; dioxin-inducible P3-450; flavoprotein-linked monooxygenase; Hydroperoxy icosatetraenoate dehydratase; microsomal monooxygenase; P1-450; P3-450; P-448; P450 form 4; P450(PA); P450-1; P450-3; P450-C; P-450d; P450-D; P450DX; P450form6; P-450MC; P450MT2; RATCYPD45; xenobiotic monooxygenase
Gene Symbols CYP1A1, CYP1A2
Host Species Mouse
Immunogen 3-methylcholanthrene induced rat cytochrome P450 protein.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13076, 13077, 1543, 1544, 24296, 24297
Target Species Human, Mouse, Monkey, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.