Learn More
CYP20A1 Antibody (Z27-P3H7*E3), DyLight 680, R&D Systems™

Description
Specifications
Specifications
| Antigen | CYP20A1 |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | Z27-P3H7*E3 |
| Conjugate | DyLight 680 |
| Dilution | Western Blot, ELISA, Immunohistochemistry |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | CYP-M, cytochrome P450 20A1, cytochrome P450 monooxygenase, cytochrome P450, family 20, subfamily A, polypeptide 1, EC 1.14, MGC22229 |
| Gene Symbols | CYP20A1 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.