Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP26A1 Monoclonal Antibody (F27 P6 A1)
GREENER_CHOICE

Catalog No. 01674090
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-090 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-090 Supplier Invitrogen™ Supplier No. MA112889
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

A suggested positive control for this product is liposarcoma cells.

CYP26A1 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This endoplasmic reticulum protein acts on retinoids, including all-trans-retinoic acid (RA), with both 4-hydroxylation and 18-hydroxylation activities. This enzyme regulates the cellular level of retinoic acid which is involved in regulation of gene expression in both embryonic and adult tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP26A1
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone F27 P6 A1
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide
Gene CYP26A1
Gene Accession No. O43174
Gene Alias CP26; Cyp26; CYP26A1; Cytochrome P450 26A1; cytochrome P450 family 26 subfamily A member 1; cytochrome P450 retinoic acid-inactivating 1; cytochrome P450, 26, retinoic acid; cytochrome P450, 26, retinoic acid A1; cytochrome P450, family 26, subfamily A, polypeptide 1; cytochrome P450, subfamily XXVIA, polypeptide 1; cytochrome P450RAI; hP450RAI; P450, retinoic acid-inactivating, 1; P450RA; P450RAI; P450RAI1; RAH; Retinoic acid 4-hydroxylase; retinoic acid hydrolase; retinoic acid hydroxylase; retinoic acid-metabolizing cytochrome
Gene Symbols CYP26A1
Host Species Mouse
Immunogen Synthetic peptide corresponding to human CYP450 26A1.
Purification Method Protein A/G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1592
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.