Learn More
Description
CYP26A1 Polyclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | CYP26A1 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | CYP26A1 |
| Gene Accession No. | O43174, O55127 |
| Gene Alias | CP26; Cyp26; CYP26A1; Cytochrome P450 26A1; cytochrome P450 family 26 subfamily A member 1; cytochrome P450 retinoic acid-inactivating 1; cytochrome P450, 26, retinoic acid; cytochrome P450, 26, retinoic acid A1; cytochrome P450, family 26, subfamily A, polypeptide 1; cytochrome P450, subfamily XXVIA, polypeptide 1; cytochrome P450RAI; hP450RAI; P450, retinoic acid-inactivating, 1; P450RA; P450RAI; P450RAI1; RAH; Retinoic acid 4-hydroxylase; retinoic acid hydrolase; retinoic acid hydroxylase; retinoic acid-metabolizing cytochrome |
| Gene Symbols | CYP26A1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
