Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP2B6 Monoclonal Antibody (OTI3D5)
GREENER_CHOICE

Catalog No. PIMA525882
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525882 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525882 Supplier Invitrogen™ Supplier No. MA525882
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CYP2B6 Monoclonal Antibody for Western Blot, IHC (P), ICC/IF, Flow

This gene, CYP2B6, encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and its expression is induced by phenobarbital. The enzyme is known to metabolize some xenobiotics, such as the anti-cancer drugs cyclophosphamide and ifosphamide. Transcript variants for this gene have been described; however, it has not been resolved whether these transcripts are in fact produced by this gene or by a closely related pseudogene, CYP2B7. Both the gene and the pseudogene are located in the middle of a CYP2A pseudogene found in a large cluster of cytochrome P450 genes from the CYP2A, CYP2B and CYP2F subfamilies on chromosome 19q.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP2B6
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI3D5
Concentration 1.00 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CYP2B6
Gene Accession No. P13107, P20813, P24460
Gene Alias 1,4-cineole 2-exo-monooxygenase; CPB6; CYP2B; CYP2B11; Cyp2b3; Cyp2b-3; CYP2B30; CYP2B6; CYP2B7; CYP2B7P; CYPIIB11; CYPIIB3; CYPIIB6; cytochrome P450 2B11; Cytochrome P450 2B3; cytochrome P450 2B6; cytochrome P450 CYP2B30; cytochrome P450 family 2 subfamily B member 6; cytochrome P-450 IIB; cytochrome P450 IIB1; Cytochrome P450 PBD-2; cytochrome P450, family 2, subfamily B, polypeptide 11; cytochrome P450, family 2, subfamily b, polypeptide 3; cytochrome P450, family 2, subfamily B, polypeptide 6; cytochrome P450, subfamily IIB (phenobarbital-inducible), polypeptide 6; cytochrome P450IIB3; Cytochrome-P450-2B11; EFVM; IIB1; P450
Gene Symbols Cyp2b3, CYP2B6
Host Species Mouse
Immunogen Full length human recombinant protein of CYP2B6 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1555, 286953, 474177
Target Species Canine, Human, Mouse, Monkey, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.