Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP2E1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA535351
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535351 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535351 Supplier Invitrogen™ Supplier No. PA535351
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CYP2E1 Polyclonal Antibody for Western Blot, IHC (P), Flow

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and is induced by ethanol, the diabetic state, and starvation. The enzyme metabolizes both endogenous substrates, such as ethanol, acetone, and acetal, as well as exogenous substrates including benzene, carbon tetrachloride, ethylene glycol, and nitrosamines which are premutagens found in cigarette smoke. Due to its many substrates, this enzyme may be involved in such varied processes as gluconeogenesis, hepatic cirrhosis, diabetes, and cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP2E1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Cyp2e1
Gene Accession No. P05181
Gene Alias 4-nitrophenol 2-hydroxylase; CP2E1; CPE1; Cyp2e; Cyp2e1; Cyp2e-1; CYPIIE1; cytochrome P450 2E1; cytochrome P450 family 2 subfamily E member 1; cytochrome P450 subfamily 2e1 (ethanol-inducible); cytochrome P450, 2e1, ethanol inducible; cytochrome P450, family 2, subfamily e, polypeptide 1; cytochrome P450, subfamily 2E, polypeptide 1; cytochrome P450, subfamily IIE (ethanol-inducible), polypeptide 1; cytochrome P450-ALC; Cytochrome P450-J; cytochrome P450RLM6; flavoprotein-linked monooxygenase; microsomal monooxygenase; P450C2E; P450-J; P450RLM6; xenobiotic monooxygenase
Gene Symbols Cyp2e1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 402-429 amino acids from the C-terminal region of human CYP2E1.
Purification Method Ammonium sulfate precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1571
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.