Learn More
CYP2E1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Shop All Proteintech Group Inc Products

Description
CYP2E1 can be detected as 50-57 kDa (full-length) and ∽40 kDa (N-terminal signal peptide cleaved). The antibody is specific to CYP2E1.
Specifications
Specifications
| Antigen | CYP2E1 |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.61 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | CYP2E1 |
| Gene Accession No. | P05181, P05182, Q05421 |
| Gene Alias | 4 nitrophenol 2 hydroxylase, CPE1, CYP2E, CYP2E1, CYP-2E1, CYPIIE1, Cytochrome P450 2E1, Cytochrome P450 J, P450 J, P450C2E |
| Gene Symbols | Cyp2e1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.