Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP2F1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103385
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103385 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103385 Supplier Invitrogen™ Supplier No. PA5103385
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Cytochrome P450 2F1.

CYP2F1 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and is known to dehydrogenate 3-methylindole, an endogenous toxin derived from the fermentation of tryptophan, as well as xenobiotic substrates such as naphthalene and ethoxycoumarin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP2F1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CYP2F1
Gene Accession No. P24903
Gene Alias C2F1; CYP2F; Cyp2f1; Cyp2f2; Cyp2f4; CYP4502F4; CYPIIF1; CYPIIF2; Cytochrome P450 2F1; cytochrome P450 2F2; cytochrome P450 family 2 subfamily F member 1; cytochrome P450, family 2, subfamily F, polypeptide 1; cytochrome P450, family 2, subfamily f, polypeptide 2; cytochrome P450, family 2, subfamily f, polypeptide 4; cytochrome P450, subfamily 2F, polypeptide 1; cytochrome P450, subfamily IIF, polypeptide 1; cytochrome P450-NAH-2; flavoprotein-linked monooxygenase; microsomal monooxygenase; naphthalene dehydrogenase; Naphthalene hydroxylase; P450-NAH-2; xenobiotic monooxygenase
Gene Symbols CYP2F1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CYP2F1(Accession P24903), corresponding to amino acid residues T451-R491.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1572
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.