Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP2S1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540656
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540656 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540656 Supplier Invitrogen™ Supplier No. PA540656
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MKYPHVQKWV REELNRELGA GQAPSLGDRT RLPYTDAVLH EAQRLLALVP Sequence homology: Cow: 93%; Dog: 93%; Goat: 83%; Guinea Pig: 82%; Horse: 82%; Human: 100%; Mouse: 90%; Pig: 93%; Rabbit: 93%; Rat: 93%; Sheep: 83%.

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum. In rodents, the homologous protein has been shown to metabolize certain carcinogens; however, the specific function of the human protein has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP2S1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CYP2S1
Gene Accession No. Q96SQ9
Gene Alias 1200011C15Rik; AU041727; C79779; CYP2S1; CYPIIS1; cytochrome P450 2S1; cytochrome P450 family 2 subfamily S member 1; cytochrome P450, 2s1; cytochrome P450, family 2, subfamily S, polypeptide 1; cytochrome P540, subfamily IIS, polypeptide 1; Hydroperoxy icosatetraenoate dehydratase; Thromboxane-A synthase; UNQ891/PRO1906
Gene Symbols CYP2S1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human CYP2S1 (aa 319-368).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29785
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.