Learn More
Description
MA3-033 detects Cytochrome P450 3A5 from HEK293 and HepG2 human cell lines, and from bovine liver and pancreas samples. MA3-033 has been successfully used in Western blot applications. By Western blot, this antibody detects a ~54 kDa band representing Cytochrome P450 3A5. The MA3-033 immunogen is a synthetic peptide corresponding to residues E S R D G T L S G E, conjugated to ovalbumin.
Specifications
Specifications
| Antigen | CYP3A5 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | F18 P3 B6 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Ascites with 0.05% sodium azide |
| Gene | CYP3A5 |
| Gene Accession No. | P20815 |
| Gene Alias | aryl hydrocarbon hydroxylase; CP35; CYP3A28; CYP3A4; CYP3A5; CYPIIIA28; CYPIIIA5; cytochrome P450; cytochrome P450 3A28; cytochrome P450 3A5; cytochrome P450 family 3 subfamily A member 5; cytochrome P450 HLp2; cytochrome P450, family 3, subfamily A, polypeptide 5; cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 4; cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 5; cytochrome P450-PCN3; flavoprotein-linked monooxygenase; HLp2; microsomal monooxygenase; P450PCN3; P450-PCN3; PCN3; Unknown (protein for MGC:127840); xenobiotic monooxygenase |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
