Learn More
Description
PA3-033 detects cytochrome P450 4A (isoforms 1, 2, & 3) from human, feline, rabbit and rat tissues. PA3-033 has been successfully used in Western blot, ICC/IF and IHC procedures. By Western blot, this antibody detects an ~50 kDa protein representing P450 4A in rat liver extract. Immunohistochemical staining of P450 4A in rat brain with PA3-033 results in perivascular and surrounding cortex staining. The PA3-033 immunogen is a synthetic peptide corresponding to residues D(431) P S R F A P D S P R H S H S(445) of rat Cytochrome P450 4A.
Specifications
Specifications
| Antigen | Cyp4a10 |
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Whole serum, PBS with 0.05% sodium azide |
| Gene | Cyp4a1 |
| Gene Accession No. | P08516 |
| Gene Alias | CYP; Cyp4a1; Cyp4a-1; Cyp4a10; Cyp4a22; CYPIVA10; cytochrome P450 4A10; cytochrome P450, 4A1; cytochrome P450, 4a10; cytochrome P450, family 4, subfamily a, polypeptide 1; cytochrome P450, family 4, subfamily a, polypeptide 10; cytochrome P450, family 4, subfamily A, polypeptide 22; cytochrome P450, IVA1; cytochrome P450-LA-omega 1; Cytochrome P452; lauric acid omega-hydroxylase; Long-chain fatty acid omega-monooxygenase; P450 |
| Gene Symbols | Cyp4a1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
