Learn More
Aviva Systems Biology Corp CYP4A22 Antibody - N-terminal region (ARP41861_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41861_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CYP4A22. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human CYP4A22. Alternate names: RP1-18D14.1ApplicationIHC, WB. Species reactivity: Cow, Human, Rat
Catalog No. 50-014-58020
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.