Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP4F22 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598504 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598504 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598504 Supplier Invitrogen™ Supplier No. PA598504
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

CYP4F22 Polyclonal Antibody for Western Blot, ELISA

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This gene is part of a cluster of cytochrome P450 genes on chromosome 19 and encodes an enzyme thought to play a role in the 12(R)-lipoxygenase pathway. Mutations in this gene are the cause of ichthyosis lamellar type 3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP4F22
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CYP4F22
Gene Accession No. Q6NT55
Gene Alias ARCI5; CYP4F22; Cytochrome P450 4F22; cytochrome P450 family 4 subfamily F member 22; cytochrome P450, family 2, subfamily E, polypeptide 2 homolog; cytochrome P450, family 4, subfamily F, polypeptide 22; INLNE; LI3
Gene Symbols CYP4F22
Host Species Rabbit
Immunogen Recombinant Human Cytochrome P450 4F22 protein (413-507AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126410
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.