Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP4V2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109613
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109613 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109613 Supplier Invitrogen™ Supplier No. PA5109613
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CYP4V2 Polyclonal Antibody for ELISA, ICC/IF, Western Blot

This gene encodes a member of the cytochrome P450 hemethiolate protein superfamily which are involved in oxidizing various substrates in the metabolic pathway. It is implicated in the metabolism of fatty acid precursors into n-3 polyunsaturated fatty acids. Mutations in this gene result in Bietti crystalline corneoretinal dystrophy. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP4V2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.89 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene CYP4V2
Gene Accession No. Q6ZWL3
Gene Alias BCD; Cy4v2; CYP4AH1; CYP4V2; Cyp4v3; Cytochrome P450 4V2; cytochrome P450 family 4 subfamily V member 2; cytochrome P450, family 4, subfamily V, polypeptide 2; docosahexaenoic acid omega-hydroxylase CYP4V2
Gene Symbols CYP4V2
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285440
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.