Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP7A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA515216
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA515216 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA515216 Supplier Invitrogen™ Supplier No. PA515216
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with rabbit based on sequence homology.

CYP7A1 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This endoplasmic reticulum membrane protein catalyzes the first reaction in the cholesterol catabolic pathway in the liver, which converts cholesterol to bile acids. This reaction is the rate limiting step and the major site of regulation of bile acid synthesis, which is the primary mechanism for the removal of cholesterol from the body.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP7A1
Applications Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Cyp7a1
Gene Accession No. P22680
Gene Alias 24-hydroxycholesterol 7-alpha-hydroxylase; CHAP; cholesterol 7 alpha hydroxylase; cholesterol 7-alpha hydroxylase; cholesterol 7-alpha-hydroxylase; Cholesterol 7-alpha-monooxygenase; CP7A; CYP7; Cyp7a1; CYP7S1; CYPVII; cytochrome P450 (cholesterol hydroxylase 7 alpha); cytochrome P450 7A1; cytochrome P450 family 7 subfamily A member 1; cytochrome P450, 7a1; cytochrome P450, family 7, subfamily a, polypeptide 1; cytochrome P450, subfamily VIIA polypeptide 1
Gene Symbols Cyp7a1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 476-504 amino acids from the C-terminal region of human CYP7A1.
Purification Method Size-exclusion - Dialysis, Ammonium sulfate precipitation
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1581
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.