Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Cypermethrin, Synthetic Pyrethroid, >98% Purity, Solid, 100 mg

Catalog No. 502706965
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6965 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6965 Supplier Abcam Supplier No. AB141018100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Synthetic pyrethroid that acts as a neurotoxin by interacting with sodium channels and inhibiting calcineurin.

  • Purity: >98%
  • Molecular Weight: 416.3 Da
  • CAS Number: 52315-07-8
  • Form: Solid
  • Description: Synthetic pyrethroid that interacts with sodium channels, inducing cellular hyperexcitability and neurotoxicity. Also acts as a calcineurin inhibitor.
  • Key Characteristics: Active in vivo and blood-brain barrier permeable.
  • Storage: Store at -20°C.

Specifications

Shipping Condition Ambient
Target Molecule Sodium Channels, Calcineurin
Content And Storage Store at -20°C.
Form Solid
Formulation 100% Cypermethrin
Quantity 100 mg
For Use With (Application) Neurotoxicity research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.