Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cypridina luciferase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA1182 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA1182 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA1182 Supplier Invitrogen™ Supplier No. PA1182
  • $578.35
    $461.65 / Each of 1

    Save $116.70

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Western blot analysis of PA1-182 detects a ~62 kDa protein in the supernatant of Cypridina luciferase-secreting cells. PA1-182 can be used for activity-independent verification of Cypridina luciferase expression.

The Cypridina luciferase is a 62kDa protein from the marine ostracod, Cypridina noctiluca. The bioluminescent enzyme is highly secreted into the cell culture media, allowing for live cell monitoring of reporter activity. Light output generated by the luciferase reaction can be correlated to the amount of Cypridina luciferase protein produced and used to determine the activity of the promoter driving Cypridina expression. Cypridina luciferase can be measured individually or in multiplex assays, such as the Cypridina-Firefly Luciferase Dual Assay Kit (Product # 16183). The Cypridina luciferase has been codon optimized for enhanced performance.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cypridina luciferase
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide
Gene Alias Cypridina noctiluca luciferase
Host Species Rabbit
Immunogen Purified recombinant Cypridina luciferase expressed in E. coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Luciferase
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.