Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyproheptadine hydrochloride, MP Biomedicals™

Catalog No. ICN19017083 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19017083 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19017083 Supplier MP Biomedicals, Inc Supplier No. 0219017083
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyproheptadine is a Ca2+ channel blocker; 5-HT2 serotonin receptor antagonist; antihistamine, antipruritic. It is a migraine prophylactic

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41354-29-4
Molecular Formula C42H50Cl2N2O3
Molecular Weight (g/mol) 701.77
MDL Number MFCD27967225,MFCD00012538,MFCD00012538,MFCD27967225,MFCD00242817
InChI Key ZEAUHIZSRUAMQG-UHFFFAOYSA-N
Synonym Periactin hydrochloride, 1-Methyl-4-(5-dibenzo(a, e)cycloheptatrienylidene) piperidine hydrochloride
PubChem CID 74764338
ChEBI CHEBI:650917
IUPAC Name dihydrogen bis(1-methyl-4-{tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaen-2-ylidene}piperidine) trihydrate dichloride
SMILES [H+].[H+].O.O.O.[Cl-].[Cl-].CN1CCC(CC1)=C1C2=CC=CC=C2C=CC2=CC=CC=C12.CN1CCC(CC1)=C1C2=CC=CC=C2C=CC2=CC=CC=C12

Specifications

Color White
Quantity 250 mg
Formula Weight 323.88 (anhydrous)
Physical Form Powder
Chemical Name or Material Cyproheptadine hydrochloride
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.