accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OH2OH2OClClCH3NCH3NHH

CAS RN 41354-29-4

IUPAC Name: dihydrogen bis(1-methyl-4-{tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaen-2-ylidene}piperidine) trihydrate dichloride
Synonyms: dihydrogen bis(cyproheptadine) trihydrate dichloride
Molecular Weight (g/mol): 701.77
Molecular Formula: C42H50Cl2N2O3
InChi Key: ZEAUHIZSRUAMQG-UHFFFAOYSA-N
SMILES: [H+].[H+].O.O.O.[Cl-].[Cl-].CN1CCC(CC1)=C1C2=CC=CC=C2C=CC2=CC=CC=C12.CN1CCC(CC1)=C1C2=CC=CC=C2C=CC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (4)
  • (1)
  • (10)
Percent Purity 
  • (2)
  • (4)
  • (6)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
Grade 
  • (10)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (10)
  • (4)
  • (1)
  • (1)
  • (6)
  • (2)
  • (1)
  • (4)
  • (4)
  • (1)
  • (10)
  • (2)
  • (4)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (10)
  • (2)
  • (1)