Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Cyproheptadine hydrochloride sesquihydrate

Catalog No. AC462670050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462670050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC462670050 Supplier Thermo Scientific Chemicals Supplier No. 462670050

missing translation for 'validMainframeMsg'

CAS: 41354-29-4 | C42H50Cl2N2O3 | 701.77 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41354-29-4
Molecular Formula C42H50Cl2N2O3
Molecular Weight (g/mol) 701.77
MDL Number MFCD27967225,MFCD00012538,MFCD00012538,MFCD27967225,MFCD00242817
InChI Key ZEAUHIZSRUAMQG-UHFFFAOYSA-N
IUPAC Name dihydrogen bis(1-methyl-4-{tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaen-2-ylidene}piperidine) trihydrate dichloride
SMILES [H+].[H+].O.O.O.[Cl-].[Cl-].CN1CCC(CC1)=C1C2=CC=CC=C2C=CC2=CC=CC=C12.CN1CCC(CC1)=C1C2=CC=CC=C2C=CC2=CC=CC=C12

Specifications

Melting Point 165.0°C
Color Clear Colorless to Light Yellow
Quantity 5 g
Solubility Information Solubility aq. soln.: very slightly soluble. Other solubilities: very soluble in methanol, soluble in chloroform, slightly soluble in ethanol
Formula Weight 350.88
Chemical Name or Material Cyproheptadine hydrochloride sesquihydrate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Toxic if swallowed.

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

IF ON SKIN: Wash with plenty of soap and water.

If eye irritation persists: Get medical advice/attention.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Wear protective gloves/protective clothing/eye protection/face protection.

If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.