Learn More
Cyproterone acetate, 98%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 207.0°C to 215.0°C |
| Ignition Residue | 0.1% max. |
| Loss on Drying | 0.5% max. (80°C) |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (HPLC) |
| Packaging | Glass Bottle |
| Quantity | 1 g |
| Solubility Information | Solubility in water: insoluble in water. Other solubilities: soluble in acetone, dichloromethane, methanol, slightly soluble in ethanol |
| Formula Weight | 416.94 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Harmful in contact with skin. Harmful if inhaled. Suspected of causing cancer. |
| Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. Call a POISON CENTER or doctor/physician if you feel unwell. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. |
| EINECSNumber | 207-048-3 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.