Learn More
GA International Permanent Cryo-LazrTAG™- Cryogenic Labels for Laser Printers (US Letter Size) PATENTED
Supplier:  GA International CL3T1WH
Encompass_Preferred
These thermoplastic waterproof labels are specifically designed to permanently adhere to vials, microtubes, microplates, glass tubes, freezer boxes, polycarbonate cryo boxes, and other containers that will undergo long-term deep-freeze or cryogenic storage in liquid or vapor phase liquid nitrogen, laboratory freezers, as well as transportation on dry ice. Resistant to multiple freeze-thaw cycles without fading, detaching, or wrinkling, these laser labels are provided in sheet format for printing with desktop and office laser printers, are wear-and-tear proof, will not jam printers, and resist UV and gamma irradiation.
Catalog No. NC0466932
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.