Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYSLTR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532717
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532717 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532717 Supplier Invitrogen™ Supplier No. PA532717
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gibbon, Marmoset, Horse (100%); Gorilla, Monkey, Mouse, Panda (94%); Dog, Bovine, Hamster, Elephant, Rabbit, Opossum (88%); Pig (82%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

Receptor for cysteinyl leukotrienes. The response is mediated via a G-protein that activates a phosphatidylinositol-calcium second messenger system. Stimulation by BAY u9773, a partial agonist, induces specific contractions of pulmonary veins and might also have an indirect role in the relaxation of the pulmonary vascular endothelium. The rank order of affinities for the leukotrienes is LTC4 = LTD4 >> LTE4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYSLTR2
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene CYSLTR2
Gene Accession No. Q9NS75
Gene Alias 2300001H05Rik; Cltr2; CysLT(2); Cyslt2; CysLT-2; CYSLT2R; CYSLTR2; cysteinyl leukotriene CysLT2 receptor; cysteinyl leukotriene receptor 2; GPCR21; G-protein coupled receptor GPCR21; G-protein coupled receptor HG57; HG57; hGPCR21; HPN321; KPG_011; PSEC0146; RSBPT32
Gene Symbols CYSLTR2
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from 1st extracellular domain of human CYSLTR2 / CYSLT2.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100056882, 57105
Target Species Human, Horse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.