Learn More
Abcam Cysteamine Hydrochloride, Solid, 113.61 Da, 25 g
Description
- Molecular weight: 113.61 Da
- Purity: >98%
- Form: Solid
- Composition: 100% Mercaptamine hydrochloride
- Application: Degradation product of cystine with various pharmacological activities; effective against cystinosis and as an antitumor agent; active in vivo
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Target Molecule | Cystine |
| Shipping Condition | Ambient |
| Buffer | 100% Mercaptamine hydrochloride |
| Content And Storage | Store at ambient temperature. |
| Form | Solid |
| Formulation | 100% Mercaptamine hydrochloride |
| Quantity | 25 g |
| For Use With (Application) | Pharmacology, Cystinosis treatment, Antitumor research |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.