Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Cysteamine Hydrochloride, Solid, 98% Purity, 100 g

Catalog No. 502706219
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6219 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6219 Supplier Abcam Supplier No. AB142575100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent pharmacological agent effective against cystinosis and known for its antitumor activity.

  • Molecular Weight: 113.61 Da
  • Purity: >98%
  • Form: Solid
  • Composition: 100% Mercaptamine hydrochloride
  • Key Attributes: Degradation product of cystine; exhibits various pharmacological activities; effective against cystinosis; acts as an antitumor agent; active in vivo

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Buffer 100% mercaptamine hydrochloride
Content And Storage Store at ambient temperature.
Form Solid
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Target Molecule Cystine
Formulation 100% Mercaptamine hydrochloride
Quantity 100 g
For Use With (Application) Pharmacological research, Cystinosis treatment, Antitumor study
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.