Learn More
MilliporeSigma™ Cysteine Sulfenic Acid Probe (DCP-Bio1)
Catalog No. NS12261MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
NS12261MG 1 mg
1 options
Catalog No. NS12261MG Supplier MilliporeSigma™ Supplier No. NS12261MG
Add to Cart
Add to Cart
Includes
DCP-Bio1: 1mg dry powder, stored under argon
DCP-Bio1, which consists of the sulfenic acid-reactive 3-(2,4-dioxocyclohexyl) propyl (DCP) appended to biotin, is a useful reagent for characterizing cellular cysteine sulfenic acid
  • New reagents have been developed to simplify the analysis and isolation of proteins bearing cysteine sulfenic acid
  • DCP-Bio1, which consists of the sulfenic acid-reactive 3-(2,4-dioxocyclohexyl) propyl (DCP) appended to biotin, is a useful reagent for characterizing cellular cysteine sulfenic acid 
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage -20°C
Quantity 1 mg