Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cysteine Sulfenic Acid Rabbit anti-All, Polyclonal, MilliporeSigma™

Catalog No. 072139I100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
072139I100 100 μL
072139I25UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 072139I100 Supplier MilliporeSigma Supplier No. 072139I100UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cysteine Sulfenic Acid Polyclonal antibody specifically detects Cysteine Sulfenic Acid in All samples. It is validated for Western Blotting, Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cysteine Sulfenic Acid
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal antibody in PBS with 0.05% sodium azide.
Host Species Rabbit
Immunogen KLH-conjugated cysteine sulfenic acid.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species All species
Content And Storage Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Serum
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.