Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Cysteinyl Leukotriene ELISA Kit (Colorimetric)
SDP

Highly purified and high bioactivity. Generating reliable and reproducible results.

$792.00 - $3020.00

Specifications

For Use With (Application) ELISA
Storage Requirements Store at 4°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP261288
SDP
View Documents
Novus Biologicals™
NBP261288
1 Kit
Each for $792.00
Only null left
Add to Cart
 
NB2612885KT
SDP
View Documents
Novus Biologicals™
NBP2612885KIT
5 Kit
Each for $3,020.00
Only null left
Add to Cart
 
Description

Description

The Non-species specific Cysteinyl Leukotriene ELISA Kit (Colorimetric) measures non-species specific Cysteinyl Leukotriene in culture supernatants and urine from any species.
Specifications

Specifications

ELISA
Store at 4°C
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.