Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cystine (DL- and meso- mixture) 98.0+%, TCI America™
SDP

Catalog No. C05181G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C05181G 1 g
C05185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C05181G Supplier TCI America Supplier No. C05181G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 923-32-0
Molecular Formula C6H12N2O4S2
Molecular Weight (g/mol) 240.292
MDL Number MFCD00084652
InChI Key LEVWYRKDKASIDU-UHFFFAOYSA-N
Synonym dl-cystine, cystine, d-cystine, 3,3'-disulfanediylbis 2-aminopropanoic acid, dicysteine, cystin, h-dl-cys-oh 2, .beta.,.beta.'-dithiodialanine, cystine, dl, cystine, l
PubChem CID 595
ChEBI CHEBI:17376
IUPAC Name 2-amino-3-[(2-amino-2-carboxyethyl)disulfanyl]propanoic acid
SMILES C(C(C(=O)O)N)SSCC(C(=O)O)N

Specifications

Melting Point 260°C
Color White
Quantity 1 g
Formula Weight 240.29
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Cystine (DL- and meso- mixture)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.