Learn More
STEMCELL Technologies Erastin, Size: 1 mg
SDP
Supplier:  STEMCELL Technologies 1000544
Encompass_Preferred
Erastin is a cystine transporter inhibitor that induces ferroptotic cell death in vitro (Li et al.). By blocking system Xc−, it induces iron-dependent programmed cell death that plays an important regulatory role in cancer, neurological disease, and acute kidney injury (Li et al.). It is shown to have increased activity in the presence of oncoproteins (Dolma et al.).CANCER RESEARCH· Induces cell death in cells expressing Ras and SV40 small T oncoprotein (IC50 = 1.25 - 5 μg/mL; Dolma et al.).· Inhibits cystine uptake through the cystine-glutamate antiporter (system Xc−) as measured by glutamate release assay (IC50 = 0.20 μM; Larraufie et al.).
Catalog No. 50-206-9352
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.