Learn More
Dojindo Molecular Technologies Inc Cystine Uptake Assay Kit, 100 tests, Fluorometric, Plate-based, Fluorescence, 490/535 nm
SDP
Supplier:  Dojindo Molecular Technologies Inc UP0512
Encompass
Cystine Uptake Assay Kit, 100 tests, Fluorometric, Plate-based, Fluorescence, 490/535 nm
Catalog No. NC2176289
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.