Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent BioTek Cytation 9 Cell Imaging Multimode Reader

Catalog No. BTCYT9UMWSN Shop All Agilent Technologies Inc. Products
Change view
Click to view available options
Model:
M-SN
MF-SN
MFA-SN
MFAW-SN
MFW-SN
MPW-SN
MW-SN
PW-SN
U-SN
UMW-SN
UW-SN
W-SN
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
BTCYT9UMWSN UMW-SN
BTCYT9MFASN MFA-SN
11-120-786 MFAW-SN
BTCYT9MFSN MF-SN
BTCYT9MFWSN MFW-SN
BTCYT9MPWSN MPW-SN
BTCYT9MSN M-SN
BTCYT9MWSN MW-SN
BTCYT9PWSN PW-SN
BTCYT9USN U-SN
BTCYT9UWSN UW-SN
BTCYT9WSN W-SN
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. BTCYT9UMWSN Supplier Agilent Supplier No. CYT9UMWSN
Get Up to 25% Off Cell Imaging Multimode Readers
View all promotions
Add to Cart
Add to Cart

It merges multimode microplate reading with widefield inverted and upright microscopy in a single platform.

  • Streamlined Discovery: A single, comprehensive platform that covers a diverse set of biochemical and cell-based assays.
  • Richer Cellular Insights: Capture more data per well with an expanded image cube capacity and enhanced optics.
  • Optimized Workflow Speed: Experience faster imaging throughput to move from raw samples to analyzed results more efficiently.
  • Superior Kinetic Data: Software-controlled gas modules create a stable, monitored environment for live cell studies.
  • Ready for Any Assay: The modular design grows with your lab, allowing you to add imaging modes as needs evolve.
  • This comprehensive system allows researchers to cover a diverse set of applications, from routine biochemical assays to complex cell-based studies.
TRUSTED_SUSTAINABILITY

Specifications

Type Cell Imaging Multi-Mode Microplate Reader
Camera Sony CMOS, 16-bit Gray Scale, Standard or WFOV
Includes Automated Upright Microscopy in Reflected and Transmitted Brightfield and Automated Widefield Microscopy in Fluorescence, Brightfield, Color Brightfield, and High Contrast Brightfield. Variable Bandwidth Monochromator Detection of Fluorescence Intensity, UV-Vis Absorbance, and Luminescence. Incubation to 45°C and Shaking.
Model UMW-SN
Magnification 1.25x to 60x
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.