Learn More
Selleck Chemical LLC Regorafenib S1178-100mg
SDP
Supplier:  Selleck Chemical LLC S1178100MG
Encompass_Preferred
Regorafenib is a multi-target inhibitor for VEGFR1 VEGFR2 VEGFR3 PDGFR Kit (c-Kit) RET (c-RET) and Raf-1 with IC50 of 13 nM/4 2 nM/46 nM 22 nM 7 nM 1 5 nM and 2 5 nM in cell-free assays respectively Regorafenib induces autophagy
Catalog No. 50-310-8143
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.