Learn More
Apexbio Technology LLC Cytidine 5-triphosphate 65-47-4 500mg
Supplier: Apexbio Technology LLC B7880500
Cytidine 5 -triphosphate (CTP CAS 65-47-4) is a pyrimidine nucleoside triphosphate that serves as a substrate in various biochemical pathways notably as a precursor for RNA synthesis and as a cofactor in phospholipid and glycoprotein biosynthesis CTP participates in the rate-limiting conversion of phosphocholine to CDP-choline catalyzed by CTP phosphocholine cytidylyltransferase a critical step in phosphatidylcholine production This process is implicated in membrane biogenesis and the provision of substrates for cell growth Additionally CTP is utilized by N-acylneuraminate cytidylyltransferase to form CMP-Neu5Ac an essential intermediate in sialylation CTP is widely employed in studies of nucleotide metabolism membrane synthesis and glycosylation pathways
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.