Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Cytidine-5'-triphosphate disodium salt, 97%

Catalog No. AAJ6223803
Change view
Click to view available options
Quantity:
500 mg
1 g
2 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6223803 1 g
AAJ62238ME 500 mg
AAJ6223804 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6223803 Supplier Thermo Scientific Chemicals Supplier No. J6223803
Add to Cart

CAS: 36051-68-0 | C9H14N3Na2O14P3 | 527.12 g/mol

Cytidine-5'-triphosphate disodium salt is a cation-permeable ligand gated ion channel (P2X purinergic receptor) agonist. It is also used as an organic intermediate, pharmaceutical intermediate, food additive and flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cytidine-5′-triphosphate disodium salt is a cation-permeable ligand gated ion channel (P2X purinergic receptor) agonist. It is also used as an organic intermediate, pharmaceutical intermediate, food additive and flavoring agent.

Solubility
Soluble in water. Insoluble in ethanol, chloroform and ether.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36051-68-0
Molecular Formula C9H14N3Na2O14P3
Molecular Weight (g/mol) 527.12
MDL Number MFCD00078193
InChI Key NFQMDTRPCFJJND-WFIJOQBCSA-L
Synonym CTP
PubChem CID 131675548
IUPAC Name disodium;(2R,3R,4S,5R)-2-(4-amino-2-oxopyrimidin-1-yl)-5-[[hydroxy-[hydroxy(phosphonooxy)phosphoryl]oxyphosphoryl]oxymethyl]oxolane-3,4-diolate
SMILES [Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]1O

Specifications

Color White
Flash Point 113°C (235°F)
Quantity 1 g
Solubility Information Soluble in water. Insoluble in ethanol,chloroform and ether.
Formula Weight 527.12
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Cytidine-5'-triphosphate disodium salt
EINECSNumber 252-849-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.