Learn More
Thermo Scientific Chemicals Cytidine-5'-triphosphate disodium salt, 97%
Description
Cytidine-5'-triphosphate disodium salt is a cation-permeable ligand gated ion channel (P2X purinergic receptor) agonist. It is also used as an organic intermediate, pharmaceutical intermediate, food additive and flavoring agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Cytidine-5′-triphosphate disodium salt is a cation-permeable ligand gated ion channel (P2X purinergic receptor) agonist. It is also used as an organic intermediate, pharmaceutical intermediate, food additive and flavoring agent.
Solubility
Soluble in water. Insoluble in ethanol, chloroform and ether.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Color | White |
| Flash Point | 113°C (235°F) |
| Quantity | 2 g |
| Solubility Information | Soluble in water. Insoluble in ethanol,chloroform and ether. |
| Formula Weight | 527.12 |
| Percent Purity | ≥98% |
| Physical Form | Powder |
| Chemical Name or Material | Cytidine-5'-triphosphate disodium salt |
Safety and Handling
| EINECSNumber | 252-849-3 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.