Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytidine, free base, 99.8%, MP Biomedicals™

Catalog No. ICN10145980 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10145980 100 g
ICN10145905 5 g
ICN10145910 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10145980 Supplier MP Biomedicals, Inc Supplier No. 0210145980
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 65-46-3 | Formula: C9H13N3O5 | Weight: 243.22 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65-46-3
Molecular Formula C9H13N3O5
Molecular Weight (g/mol) 243.22
InChI Key UHDGCWIWMRVCDJ-UHFFFAOYNA-N
Synonym cytidine, cytosine riboside, 1-beta-d-ribofuranosylcytosine, 1beta-ribofuranosylcytosine, 4-amino-1-beta-d-ribofuranosyl-2 1h-pyrimidinone, beta-d-ribofuranoside, cytosine-1, 1-beta-ribofuranosylcytosine, cytidin, zytidin, 4-amino-1beta-d-ribofuranosyl-2 1h-pyrimidinone
PubChem CID 6175
ChEBI CHEBI:17562
IUPAC Name 4-amino-1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one
SMILES NC1=NC(=O)N(C=C1)C1OC(CO)C(O)C1O

Specifications

Color White
Assay Percent Range 99.8%
Quantity 100 g
Formula Weight 243.22
Percent Purity 99.8%
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material Cytidine, free base
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.