Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytidine, MP Biomedicals™

Catalog No. MP219465101 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219465101 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219465101 Supplier MP Biomedicals, Inc Supplier No. 0219465101
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Composed of the base cytosine linked to the five-carbon sugar d-ribose. Cytidine, MP Biomedicals is a pyrimidine nucleoside with a variety of analogs that display potentially useful pharmacology.

Cytidine, when intraperitoneal administered in combination with choline, improves nerve regeneration and functional recovery in a rat model of sciatic nerve injury. It is also used as an ex vivo drug selectable marker in gene-modified primary bone marrow stromal cells for cytidine deaminase assay.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65-46-3
Molecular Formula C9H13N3O5
Molecular Weight (g/mol) 243.22
InChI Key UHDGCWIWMRVCDJ-UHFFFAOYNA-N
Synonym cytidine, cytosine riboside, 1-beta-d-ribofuranosylcytosine, 1beta-ribofuranosylcytosine, 4-amino-1-beta-d-ribofuranosyl-2 1h-pyrimidinone, beta-d-ribofuranoside, cytosine-1, 1-beta-ribofuranosylcytosine, cytidin, zytidin, 4-amino-1beta-d-ribofuranosyl-2 1h-pyrimidinone
PubChem CID 6175
ChEBI CHEBI:17562
IUPAC Name 4-amino-1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one
SMILES NC1=NC(=O)N(C=C1)C1OC(CO)C(O)C1O

Specifications

Absorbance λ max (water): 270-272nm
Melting Point 208°C to 218°C
Quantity 1 g
Solubility Information Soluble in water (50mg/mL, clear colorless to faint yellow solution).
Formula Weight 243.2
Grade Cell Culture Reagent
Physical Form Powder
Recommended Storage Store at Room Temperature (15-30°C), dessicate.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.