Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYTL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101322
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101322 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101322 Supplier Invitrogen™ Supplier No. PA5101322
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CYTL1.

C17 is a cytokine-like protein specifically expressed in bone marrow and cord blood mononuclear cells that bear the CD34 surface marker (Liu et al., 2000 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYTL1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CYTL1
Gene Accession No. Q9NRR1
Gene Alias 4930443F05Rik; 4Cytl1; C17; C4orf4; Cyt1; CYTL1; cytokine like 1; cytokine-like 1; cytokine-like protein 1; cytokine-like protein C17; Gm147; Protein C17; RGD1561846; UNQ1942/PRO4425
Gene Symbols CYTL1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CYTL1(Accession Q9NRR1), corresponding to amino acid residues Y68-C118.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 231162, 498392, 54360
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.