Learn More
Cayman Chemical 25I-NBFhydrochlorIde 5mg
Supplier: Cayman Chemical 140665 MG
A cGMP phosphodiesterase inhibitor (IC50s 0 5-0 76 and 0 15 UM for PDE5 and PDE6 respectively) selective for PDE5 and PDE6 over PDE9 PDE10 and PDE11 (IC50s 35 22 and 11-33 UM respectively) enhances the vasodilatory effects of NO in a range of vascular tissues by prolonging the cGMP-mediated activation of cGMP-dependent protein kinase activates both rat and human GPR35 (EC50s 16 nM and 0 84 UM respectively)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.