Learn More
AdipoGen Cytochalasin B (1 mg)
SDP
Supplier:  AdipoGen AGCN20504M001
Encompass_Preferred
Cytochalasin B (1 mg), AdipoGen Life Sciences. Chemical. CAS: 14930-96-2. Formula: C29H37NO5. MW: 479.6. Isolated from Phoma sp. Cell permeable mycotoxin. Actin polymerization inhibitor. Binds to the barbed end of actin, reversibly inhibiting the elongation and shortening of actin filaments. Induces nuclear extrusion. By disrupting actin polymerization, blocks diverse cellular functions, including cell division, migration, phagocytosis, exocytosis and chemotaxis. Induces DNA fragmentation. Used in cytoskeletal reorganization, cell imaging and organelle trafficking studies. Potent antitumor activity. Causes cell cycle arrest and induces apoptosis in cancer cells.
Catalog No. 50-149-0009