Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome b-245/p22phox Mouse anti-Human, Clone: CS9, MilliporeSigma™

Catalog No. MABS2191100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS2191100 100 μg
MABS219125 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABS2191100 Supplier MilliporeSigma Supplier No. MABS2191100UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytochrome b-245/p22phox Monoclonal antibody specifically detects Cytochrome b-245/p22phox in Human samples. It is validated for Immunoprecipitation, Inhibition, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytochrome b-245/p22phox
Applications Immunoprecipitation, Inhibition Assays, Western Blot
Classification Monoclonal
Clone CS9
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in PBS without azide.
Host Species Mouse
Immunogen Partially purified human neutrophil flavocytochrome b.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.