Learn More
Invitrogen™ Cytochrome C Monoclonal Antibody (6H2.B4)

Description
This antibody reacts with native Cytochrome C, but does not recognize denatured cytochrome C. The antibody recognizes a region in the vicinity of amino acid 62 of rat Cytochrome C.
Specifications
Specifications
| Antigen | Cytochrome C |
| Applications | ELISA, Immunoprecipitation, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 6H2.B4 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.4 |
| Gene | Cycs |
| Gene Accession No. | P62897, P62898, P84029, P99999 |
| Gene Alias | APOCYTOCHROME C-A; AT1G22840; AT4G10040; ATCYTC-A; CG17903; CG17903-PA; CG17903-PB; CRF; CYC; CYC2; Cycs; CYCSA; Cyct-c-p; Cyt c; cyt c-p; cyt.c; CytC; Cyt-c; CYTC-1; CYTC2; Cyt-c2; CytC-2; cyt-c-p; cytC-p; Cyt-c-p-PA; Cyt-c-p-PB; Cytochrome c; cytochrome c DC4; Cytochrome c proximal; Cytochrome C, expressed in somatic tissues; cytochrome c, somatic; CYTOCHROME C-1; CYTOCHROME C-2; CYTOCHROME C-A; cytochrome c-like; cytochrome c-like protein; Cytochrome c-proximal; cytochrome-c-p; Cytochrome-c-proximal; DC4; dCyt.C-p; DMc01; Dmel\CG17903; Dmel_CG17903; F19G10.20; F19G10_20; hc1; HCS; l(2)k13905; lethal (2) k13905; LOC100341175; LOC100357840; MGC128356 protein; pDMc01; RH17228p; T5L19.170; T5L19_170; THC4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.