Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytochrome C Monoclonal Antibody (37BA11)
GREENER_CHOICE

Catalog No. 456100
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-610-0 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-610-0 Supplier Invitrogen™ Supplier No. 456100
  • $582.00
    $436.65 / Each of 1

    Save $145.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

45-6100 was used successfully in the western blot analysis of cytochrome C using human breast cancer cell line lysates. Positive control: Isolated mitochondria from Human, Bovine, Rat and Mouse heart; Human fibroblasts; HeLa cells. WB: Predicted molecular weight: 14 kDa. ICC/IF: Heat induced antigen retrieval in 0.1 M Tris-HCl, 5% urea, pH 9.5 for 5 min at 95°C improves signal.

Cytochrome C is an electron transporting protein that resides within the intermembrane space of the mitochondria, where it plays a critical role in oxidative phosphorylation and the production of cellular ATP. Release of cytochrome C from the mitochondria to the cytosol is triggered by apoptotic stimuli. In the cytoplasm, the cytochrome C binds apoptotic protease activating factor which activates the apoptotic initiator procaspase 9. Cytosolic cytochrome C functions in the activation of caspase 3, an ICE family molecule that is a key effector of apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytochrome C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 37BA11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation HEPES buffered saline with 0.02% sodium azide
Gene Cycs
Gene Accession No. P62894, P62897, P62898, P99999
Gene Alias CG17903; CG17903-PA; CG17903-PB; CRF; CYC; CYC2; Cycs; CYCSA; Cyct-c-p; Cyt c; cyt c-p; cyt.c; CytC; Cyt-c; CYTC2; Cyt-c2; CytC-2; cyt-c-p; cytC-p; Cyt-c-p-PA; Cyt-c-p-PB; Cytochrome c; cytochrome c DC4; Cytochrome c proximal; Cytochrome C, expressed in somatic tissues; cytochrome c, somatic; Cytochrome c-2; cytochrome c-like; cytochrome c-like protein; Cytochrome c-proximal; cytochrome-c-p; Cytochrome-c-proximal; DC4; dCyt.C-p; DMc01; Dmel\CG17903; Dmel_CG17903; hc1; HCS; l(2)k13905; lethal (2) k13905; LOC100341175; LOC100357840; MGC128356 protein; pDMc01; RH17228p; THC4
Gene Symbols Cycs
Host Species Mouse
Immunogen Bovine heart Cytochrome C.
Purification Method IgG fraction
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13063, 25309, 510767, 54205
Target Species Human, Mouse, Rat, Bovine, C. elegans
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.