Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Cytochrome c Native Protein
SDP

Catalog No. NB002026 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
0.1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002026 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002026 Supplier Novus Biologicals™ Supplier No. NBP1991810.1G

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Highly purified. Generating reliable and reproducible results.

Native Cytochrome c The Cytochrome c Native Protein is derived from Bovine heart. The Cytochrome c Native Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration Lyoph
For Use With (Application) SDS-PAGE
Formulation Lyophilized.
Gene ID (Entrez) 54205
Molecular Weight (g/mol) 12.327 kDa
Quantity 0.1 g
Source Bovine heart
Storage Requirements Store at -20 to -80C. Avoid freeze-thaw cycles.
Gene Alias CYCHCS, cytochrome c, cytochrome c, somatic, THC4
Gene Symbol CYCS
Reconstitution Reconstitute in H2O to a concentration of 10 mg/mL.
Buffer Lyophilized.
Purity or Quality Grade >97%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.