Learn More
Description
Specifications
Specifications
| Concentration | 5mg/mL |
| For Use With (Application) | Bioactivity |
| Formulation | 25mM Tris-HCl buffer, pH 7.8, 5mM EDTA, and 39mM n-dodecyl beta-D-maltoside |
| Quantity | 250 μg |
| Source | Bovine |
| Immunogen | Cytochrome C Oxidase |
| Storage Requirements | Store at -20°C. Avoid freeze-thaw cycles. |
| Regulatory Status | RUO |
| Common Name | Cytochrome c Oxidase/Complex IV consists of 7-13 different proteins and is isolated as a dimer of 250kDa of which heme a is an integral part. |
| Conjugate | Unconjugated |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
