Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP1A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112867
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112867 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112867 Supplier Invitrogen™ Supplier No. PA5112867
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CYP1A1 Polyclonal Antibody for Western Blot, ELISA

This gene, CYP1A1, encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and its expression is induced by some polycyclic aromatic hydrocarbons (PAHs), some of which are found in cigarette smoke. The enzyme's endogenous substrate is unknown; however, it is able to metabolize some PAHs to carcinogenic intermediates. The gene has been associated with lung cancer risk. A related family member, CYP1A2, is located approximately 25 kb away from CYP1A1 on chromosome 15.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP1A1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4.56 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Alias AHH; AHRR; aromatic compound inducible; aryl hydrocarbon hydroxylase; CP11; CYP; CYP1; Cyp1a1; Cyp1a-1; Cyp1a2; Cyp45c; Cypc45c; CYPIA1; cytochrome P1-450; cytochrome P1-450, dioxin-inducible; cytochrome P450 1A1; cytochrome P450 family 1 subfamily A member 1; cytochrome P450 family 1 subfamily a polypeptide 2; cytochrome P450 form 6; cytochrome P450 subfamily I (aromatic compound-inducible) member A1 (C6 form c) (C6 form c); cytochrome P450 subfamily I, polypeptide 1; cytochrome P450, 1a1; cytochrome P450, 1a1, aromatic compound inducible; cytochrome P450, family 1, subfamily a, polypeptide 1; cytochrome P450, subfamily I (aromatic compound-inducible), member A1 (C6, form c) (C6, form c); cytochrome P450, subfamily I (aromatic compound-inducible), polypeptide 1; cytochrome P450-C; cytochrome P450MT2; Cytochrome P450MT2A; Cytochrome P450MT2B; Cytochrome P450-P1; dioxin-inducible; flavoprotein-linked monooxygenase; Hydroperoxy icosatetraenoate dehydratase; microsomal monooxygenase; P1-450; P450; P450 form 6; P450-1; P450-C; P450DX; P450form6; P-450MC; P450MT2; P450-P1; xenobiotic monooxygenase
Host Species Rabbit
Immunogen Recombinant Zebrafish Cytochrome P450 1A1 protein (1-75AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Zebrafish
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.