Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytochrome P450 Reductase Monoclonal Antibody (OTI3F10)
GREENER_CHOICE

Catalog No. PIMA524977
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524977 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524977 Supplier Invitrogen™ Supplier No. MA524977
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytochrome P450 Reductase Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

This gene encodes an endoplasmic reticulum membrane oxidoreductase with an FAD-binding domain and a flavodoxin-like domain. The protein binds two cofactors, FAD and FMN, which allow it to donate electrons directly from NADPH to all microsomal P450 enzymes. Mutations in this gene have been associated with various diseases, including apparent combined P450C17 and P450C21 deficiency, amenorrhea and disordered steroidogenesis, congenital adrenal hyperplasia and Antley-Bixler syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytochrome P450 Reductase
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI3F10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene POR
Gene Accession No. P16435
Gene Alias 4933424M13Rik; CCR; CPR; CY DKFZp686G04235; CYPOR; cytochrome p450 oxidoreductase; cytochrome P450 reductase; DKFZp686G04235; FLJ26468; I79_007261; LOC101115252; NADPH Cytochrome; NADPH-cytochrome P450 oxidoreductase; NADPH-cytochrome P-450 oxidoreductase; NADPH-cytochrome P450 reductase; NADPH--cytochrome P450 reductase; NADPH-cytochrome P-450 reductase; NADPH-dependent cytochrome P450 reductase; P450 (cytochrome) oxidoreductase; P450 Reductase; P450R; Por; unnamed protein product
Gene Symbols POR
Host Species Mouse
Immunogen Full length human recombinant protein of POR produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5447
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.